Would you like to log in again?
VWR® Traceable® Flow Sensors Compatible with Hub-Sensor Monitoring System
Supplier: VWR
6641VW 6638VW 6642VW 6639VW
76403-490EA 236.12 USD InStock
76403-490 76403-492 76403-488 76403-486
VWR® Traceable® Flow Sensors Compatible with Hub-Sensor Monitoring System
Monitoring Systems Environmental Monitoring Systems
Stay in touch with all your critical environments.
  • Hub touch screen display receives and indicates all data in a central location
  • Hub links up to ten discrete BLE sensors for monitoring refrigerator/freezer temperatures, humidity/temperature, and liquid flow
  • View current conditions anywhere - Cloud-based interface sends email, text, and push notifications
  • Hub network includes Ethernet and Wi-Fi connectivity options for enhanced flexibility
  • Eliminate wires and cables currently connected to your temperature and humidity sensors
  • Connect to your data via smartphone, tablet, or PC using TraceableLIVE cloud service (requires subscription)
The Traceable® hub-sensor system offers continuous monitoring of up to ten sensors. The hub collects data from the individually calibrated sensors via Bluetooth Low Energy (BLE) while simultaneously displaying measurements. The hub is compatible with TraceableLIVE® cloud service, allowing multiple users to stay connected and monitor critical environments 24/7 wherever they go!
The hub flow sensors are available in different flow ranges and discharge configurations.
Traceable® products have an individually numbered Traceable® certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2005(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, saving you both time and money by not having to calibrate separately.
NIST-traceable A2LA calibration also helps you meet quality standards and regulatory compliance for ISO, FDA, cGMP, VFC, CAP, CLIA and Joint Commission requirements.
Certifications: NIST-traceable calibration report.
Accessories information: USB power cable.
Ordering information: TraceableLIVE® Cloud Service requires subscription.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty