Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
BRAND Pipette Holders for Transferpette® S, BrandTech
Supplier: BrandTech
Brand®
704812 704811 704807
76383-124EA 75.91 USD InStock
76383-124 76383-120 76383-122
BRAND Pipette Holders for Transferpette® S, BrandTech
Pipette Stands
Keep your Transferpette® S pipettes close at hand with these storage options.
  • Designed to fit 2020 and later Transferpette® S pipettes (serial numbers 20A and higher)
Carousel rack holds up to six pipettes. Hooks can be replaced to fit older Transferpette® S pipettes, HandyStep® touch or HandyStep® S instruments.
Shelf mount allows easy mounting of one Transferpette® S single- or multichannel instrument to a shelf or bench edge. It can also be used to retrofit older BRAND® Transferpette® S carousels to newer instruments.
Wall mount allows easy mounting of current model Transferpette® S single- or multichannel instruments to a vertical surface.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty