Would you like to log in again?
Quartz Reduction Tube, PerkinElmer
Supplier: PerkinElmer
N2411300
10138-428EA 206.73 USD InStock
10138-428
Quartz Reduction Tube, PerkinElmer
Ovens Tube Furnaces
These quartz reduction tubes are composed of pure fused quartz which is 99% SiO₂. Fused quartz has been chosen because of its very high purity and low thermal expansion rate.
All combustion tubes are pressure tested to eliminate any tubes which might contain weak spots. In addition, strict specifications are used to ensure that the combustion tubes fit in an airtight connection when placed in the analyzer.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty