Would you like to log in again?
Torch Alignment Tool for ELAN 9000/6x00/DRCs and NexION 300/350, PerkinElmer
Supplier: PerkinElmer
ELAN®
WE015554
89260-244EA 318.51 USD InStock
89260-244
Torch Alignment Tool for ELAN 9000/6x00/DRCs and NexION 300/350, PerkinElmer
Lights Flashlights
The torch alignment tool is for use with demountable quartz torches in ELAN® 9000, 6x00, DRC II, and DRC-e, and all NexION® 300 and 350 ICP-MS instruments.
This tool is used to align the torch in the load coil.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty