Would you like to log in again?
High Sensitivity GemTip Capillary Assembly, PerkinElmer
Supplier: PerkinElmer
N0371522
10138-028EA 1212.74 USD InStock
10138-028
High Sensitivity GemTip Capillary Assembly, PerkinElmer
Atomizers
This high sensitivity GemTip capillary assembly is for use with AAnalyst® 100/300/200/400/700/800 and earlier AA series instruments. Only includes capillary needle assembly, not complete assembly.
This capillary assembly is included in the GemTip high-sensitivity plastic body nebulizer complete assembly (N0370393). It is compatible with Models 5100PC/5000/3300/3110/3100 AA instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty