Would you like to log in again?
Cyclonic/Concentric Sample Introduction Torch Module for Optima™ 2x00/4x00/5x00/7x00 DV, PerkinElmer
Supplier: PerkinElmer
N0770607 N0770607
97013-832EA 5501.14 USD InStock
97013-832 CA97013-832
Cyclonic/Concentric Sample Introduction Torch Module for Optima™ 2x00/4x00/5x00/7x00 DV, PerkinElmer
Chromatography Supplies Chromatography Spare Parts
This quick-change sample introduction module, with cyclonic spray chamber and MEINHARD® concentric nebulizer technologies, includes core consumables and components for Optima™ 2x00 DV, 4x00 DV, 5x00 DV, and 7x00 DV ICP-OES series instruments.
Delivery information: This kit includes: (1) MEINHARD® Type C Concentric Glass Nebulizer, (1) 1-Slot Quartz Torch, (1) Cyclonic Spray Chamber Assembly, (1) Quick-Change Torch Module Mounting Block, (1) 2.0 mm Int.Ø Alumina Injector, (1) Nebulizer Tubing Assembly, and (1) Polyethylene Tubing (Int.Ø×L: 0.58 mm × 25.4 cm)
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty