Would you like to log in again?
UniFit Connector for SeaSpray™ and MicroMist™ Nebulizers, PerkinElmer
Supplier: PerkinElmer
N0774077
10139-150PK 218.59 USD InStock
10139-150
UniFit Connector for SeaSpray™ and MicroMist™ Nebulizers, PerkinElmer
Chromatography Supplies Chromatography Spare Parts
This UniFit Connector is designed for use with concentric glass nebulizers in Optima™ 4x00/5x00/7x00/8x00 and Avio® 200/500 ICP-OES series instruments in addition to ELAN® 5000/6x00/9000/DRCs and NexION® 300/350/1000/2000 ICP-MS series instruments.
  • UniFit connector is a convenient drain connector for glass cyclonic spray chambers
  • 700 mm overall length
  • 0.75Int.Ø.×1.3Ext.Ø mm
This assembly includes 3 UniFit connectors.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty