Would you like to log in again?
Bottle, PFA, with Cap, PerkinElmer
Supplier: PerkinElmer
N0777406 N0777407
10139-374EA 339.62 USD InStock
10139-374 10139-372
Bottle, PFA, with Cap, PerkinElmer
Bottles Plastic Bottles
PFA bottles, with cap, are available in multiple capacities and dimensions.
  • Multiple sizes available
  • Supplied with cap
PFA bottles are ideal for a wide range of demanding fluid handling and storage applications in general laboratory, semiconductor, and environmental industries.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty