Would you like to log in again?
EzyLok Kit for Glass Concentric Nebulizers, PerkinElmer
Supplier: PerkinElmer
N0777413
10139-382EA 158.46 USD InStock
10139-382
EzyLok Kit for Glass Concentric Nebulizers, PerkinElmer
Atomizers
This EzyLok kit is designed for use with Optima™ 4×00/5×00/7×00/8×00 and Avio® 200/500 ICP-OES series instruments. It is designed for use with Conikal™, slurry, SeaSpray™, and MicroMist™ nebulizers.
The EzyLok kit is included in the assembly with the Conikal™ nebulizer, 2 ml/min(10138-152), slurry nebulizer, 4 ml/min (10138-154), SeaSpray™ nebulizer, 2 ml/min(10138-174), MicroMist™ nebulizer, 0.6 ml/min (10138-176), MicroMist™ nebulizer, 0.2 ml/min (101690-960), MicroMist™ nebulizer, 0.1 ml/min (101690-962), and the MicroMist™ nebulizer, 0.05 ml/min (10138-178) for Optima 4×00/5×00/7×00/8×00 and Avio 200/500. This kit includes EzyLok connector for 4 mm tubing,  EzyLok 6 mm hose adapter, and nebulizer hose clip.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty