Would you like to log in again?
EzyLok Nebulizer Pressure Hose Clips, PerkinElmer
Supplier: PerkinElmer
N0773197
10136-330PK 91.5 USD InStock
10136-330
EzyLok Nebulizer Pressure Hose Clips, PerkinElmer
Clamps Tubing and Sanitary Clamps
Hose clips are designed for use with Avio® 200/500 and Optima™ 4×00/5×00/7×00/8×00 ICP-OES series instruments.
  • Designed for use with EzyLok tubing in SeaSpray™ and MicroMist™ nebulizers
These clips are included in the EzyLok kit for SeaSpray™ and MicroMist™ nebulizers (10139-382).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty