Would you like to log in again?
Metro Indicator Flag for WavDri™, Drying Tray, Intermetro Industries
Supplier: Intermetro
DT-KB DT-KB
76407-174PK 2.68 USD InStock
76407-174 CA76407-174
Metro Indicator Flag for WavDri™, Drying Tray, Intermetro Industries
Labels
Quickly identify if WavDri™ trays have been rotated.
Visually alternates from red to white when using the clamshell rotation feature for a consistent and documented rotation process.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty