Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Techne® Prime Pro 48 Real-Time PCR System and Accessories, Cole-Parmer®

Supplier: Cole Parmer
9394514
76407-102EA 20177.15 USD InStock
76407-102
Techne® Prime Pro 48 Real-Time PCR System and Accessories, Cole-Parmer®
The Prime Pro 48 real time PCR system is a high specification, economical real time thermal cycler that accommodates a unique 48-well polypropylene PCR plate utilising the same geometry as standard 384-well plates, but only ¹/₈ of the size.

  • Fast cycling: Runs 40 cycles in under 20 minutes when optimized
  • Process up to 40% more samples per hour than a traditional 96-well instrument
  • Up to 5x greater uniformity than any other block-based qPCR instrument
  • Uses four colors for easy multiplexing
  • Uniformity of ±0.1 °C at 95 °C

Suitable for both single labs and core facilities, the thermal cycler's fast cycling enables several experiments per day. A specially designed thermal block delivers a unique heating and cooling system that provides accurate ±0.1 °C temperature control and quickly cycles from one temperature to the next.

An advanced high-performance optical system delivers precise and sensitive fluorescence detection, facilitating all four color multiplex applications. The qPRC unit is MIQE compliant, features an adaptive LED control and high-resolution melt (HRM) functionality. Easy-to-use software provides instrument control, data collection, and data analysis. An ergonomic plate loading dock allows for quick loading of the plate and is backlit for clear visibility.

Prime Pro 48 also accepts all popular brands of reagents. Always reads all 48 wells, with all 4 filters at every cycle, and automatically normalizes fluorescence - never saturate the detector. Eliminate cross talk.

The ProDock (included) makes cell loading easier. The ergonomic platform fits into standard centrifuge rotors and is backlit for sample visualization, also works well with 16-channel pipettes.

Delivery information: Includes ProDock and Squeegee, ProControl and ProStudy software USB stick, 50 PCR plates, 50 plate seals, Ethernet cable, and power cord.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty