Would you like to log in again?
Human MS compatible human protein extract, digest
Supplier: Promega
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
V6951
PAV6951EA 661.88 USD InStock
PAV6951
Human MS compatible human protein extract, digest
Proteins and Peptides
The mass spectrometry-compatible human protein extracts are designed for monitoring mass spectrometry instrument performance. The extracts are predigested and cleaned up by solid-phase extraction for immediate use in liquid chromatography/mass spectrometry (LC-MS) analysis.
  • Extract reproducibility is assured by stringent control over the manufacturing process
  • Each protein extract is monitored by LC-MS to confirm composition and protein integrity
  • To be used primarily for instrument performance monitoring
Lot-to-lot consistency of extracts is monitored by various protein and peptide qualitative and quantitative methods, including LC-MS and amino acid analysis. Our manufacturing process assures compatibility with LC-MS by monitoring non-specific protein fragmentation, non-biological post-translational modifications and, for digested extracts, minimal undigested peptides.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty