Would you like to log in again?
Masterflex® L/S Variable-Speed Pump System, Cole-Parmer
Supplier: Cole Parmer
77715-90 77715-91
76403-128EA 2370.2 USD InStock
76403-128 76403-126
Masterflex® L/S Variable-Speed Pump System, Cole-Parmer
Pumps Peristaltic Pumps
Compact and cost effective for general fluid transfer applications.
  • Flow rate 0.42 to 2900 ml/min with L/S tubing—flow rate depends on drive rpm, pump head, and tubing size
  • Simple operation with single-turn speed control
  • Separate power switch enables you to maintain speed setting when turning drive on/off; front-panel LED indicates power on
  • Reversible motor lets you easily prime/purge tubing and pump in either direction
  • Remote control of start/stop via DB9 connector on back of drive
  • Fixed occlusion
  • Automatic tubing retention
The Masterflex L/S Variable-Speed Pump system includes a variable speed drive, Easy-Load pump head for precision tubing, and handheld remote controller to easily control you pump while filling and dispensing.
Accessories information: Drive 07555-00, pump head 07514-10, and handheld controller 07528-80.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty