Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 0.25 N volumetric solution

Supplier: VWR
Warning
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

ANAC83196-540
80721-820CS 259.09 USD InStock
80721-820
Sodium hydroxide 0.25 N volumetric solution
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1824
ADR: 8,III
Order Now
Specification Test Results
Normality 0.2490-0.2510 N
NIST traceable Confirmed

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty