Would you like to log in again?
Whatman™ Grade 597 Optima Qualitative Filter Papers, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
989410103 989410101 989410104 989410108 989410104 989410106 989410107 98949613 98949329 989410101 989410108 989410106 989410103 989410102 989410110 989410105 98949552 989410107 98949552 989410109 989410102 98949613 989410109 989410105 98949329 989410110
76402-794PK 49.8 USD InStock
76402-794 76402-796 76402-792 76402-802 CA76402-814 76402-804 CA76402-812 CA76402-798 CA76402-810 CA76402-796 76402-800 76402-806 76402-808 CA76402-816 CA76402-794 CA76402-792 CA76402-802 76402-814 CA76402-800 76402-816 76402-810 76402-798 76402-812 CA76402-808 CA76402-806 CA76402-804
Whatman™ Grade 597 Optima Qualitative Filter Papers, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
Standard Whatman grade filter paper for a variety of routine applications in food and beverage industries. Well-suited for determination of fat content in food or removal of carbon dioxide and turbidity from beverages such as beer.
  • Available in sheets and circles
  • Medium fast filtration rate
  • Minimum 90% cellulose content
  • Technical and scientific support
These cellulose filters are used in qualitative analytical techniques to determine and identify materials.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty