Would you like to log in again?
Caliber™ Type VII Gauze Sponges, DUKAL™ Corporation
Supplier: Dukal
C517 C84166 C82082 C84088 C5189 C5129 C84688 C5139 C82122 C83082 C5119 C83083
76407-990BG 3.82 USD InStock
76407-990 76407-988 76407-978 76407-984 76407-996 76407-986 76408-000 76407-998 76407-980 76407-992 76407-982 76407-994
Caliber™ Type VII Gauze Sponges, DUKAL™ Corporation
First Aid Supplies Wound Dressings
Gauze sponges are designed for use in a variety of applications, including general wound care, dressings, prepping, wound packing, and debriding.
  • 100% woven cotton
  • No natural rubber latex
  • Available in sterile and non sterile formats
Highly absorbent sponges offer exceptional clinical performance.
Certifications: Meets USP Type VII requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty