Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
MINIPULS® Connection Kits, Gilson®
Supplier: Gilson
F1825057 F1825056 F1825054
76406-976EA 169.53 USD InStock
76406-976 76406-974 76406-972
MINIPULS® Connection Kits, Gilson®
Fittings and Connectors
Connector kit accessories for the MINIPULS® 3 and Evolution peristaltic pumps come in a variety of types.
  • Metal sleeve connectors for small bore tubing
  • Connector kits for flanged tubing
  • Plastic connectors for large bore tubing
These kits contain five connectors each. The connectors are available in a three sizes to fit a variety of tubing of different inner diameters.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty