Would you like to log in again?
NanoXact Aminated Silica Shelled nanoparticles 1 mg/ml in EtOH, nanoComposix
Supplier: nanoComposix
AGAH50-30M AGAH100-10M AGAH50-10M AGAH50-1M AGAH100-5M AGAH100-1M AGAH100-30M AGAH50-5M
103819-576EA 306.92 USD InStock
103819-576 103819-574 103819-568 103819-566 103819-578 103819-572 103819-580 103819-570
NanoXact Aminated Silica Shelled nanoparticles 1 mg/ml in EtOH, nanoComposix
Beads Microcarriers
Silver nanoparticles shelled with aminated silica, versatile surface for a wide variety of applications
  • High quality nanoparticles, precisely engineered and highly characterized
  • Unagglomerated and monodisperse
  • High concentratio. Aminated silica shell offers robust and versatile surface for a wide variety of applications
Spherical silver nanoparticles, 1 mg/ml silver concentration. Aminated silica shell offers high stability in a variety of solvents and a versatile surface for attaching functional groups or creating hydrophobic & hydrophilic surfaces. Silica shell allows easier deposition onto substrates and incororation into many different solvents, resins, and curable materials because the silica shell provents metal nanoparticles from touching directly / aggregating, helping to preserve the particles' nano and optical properties.
Each batch is extensively characterized to meet highest quality standards, ensuring reliable & reproducible results. All products supplied with batch-specific COA's with TEM, DLS, Zeta, & UV-Vis. to meet the highest quality standards
Colloidal silver has applications in Diagnostics, Antimicrobial coatings, Electronics and Photovoltaics, SERS, Optics, Nanotoxicology, Plasmonics (SPR / Surface Plasmon Resonance), novel Color Enhancements and Pigments, Molecular Detection, Sensors, and Photothermal and Microwave-based Therapies.
Custom formulations and ISO13485/cGMP compliant materials can be provided on request.
Each batch is extensively characterized to meet highest quality standards, ensuring reliable & reproducible results. All products supplied with batch-specific COA's with TEM, DLS, Zeta, & UV-Vis. to meet the highest quality standards
Colloidal silver has applications in Diagnostics, Antimicrobial coatings, Electronics and Photovoltaics, SERS, Optics, Nanotoxicology, Plasmonics (SPR / Surface Plasmon Resonance), novel Color Enhancements and Pigments, Molecular Detection, Sensors, and Photothermal and Microwave-based Therapies.
Custom formulations and ISO13485/cGMP compliant materials can be provided on request.
Delivery information: Ships via FedEx Priority Overnight (Domestic). Cold packaging included.
Due to temperature-sensitive nature of these materials, they will only be shipped during the week in order to prevent products from sitting in an uncontrolled temperature environment over the weekend.
Caution: DO NOT FREEZE. Store at 2 to 8 °C and away from light when not in use to maximize shelf life. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty