Would you like to log in again?
BioPure Silver Nanospheres 1 mg/ml Carboxyl (lipoic acid) surface for conjugation, nanoComposix
Supplier: nanoComposix
AGLB50-30M AGLB100-30M
103819-664EA 4027.69 USD InStock
103819-664 103819-658
BioPure Silver Nanospheres 1 mg/ml Carboxyl (lipoic acid) surface for conjugation, nanoComposix
Beads Microcarriers
Purified, concentrated, high quality silver nps; sterile and endotoxin-free. Carboxylic acid surface.
  • High quality nanoparticles, precisely engineered and highly characterized
  • Unagglomerated and monodisperse
  • High concentration materials (1 mg/ml, 9.27 mm, 1000 ppm, ~40 to 135 OD [depending on size])
  • Extensively purified to remove residual reactants
  • Manufactured sterile; provided in ultra to pure (Milli to Q) DI water
  • BioPure mateirals ideal for in to vitro and in to vivo experiments, nanotoxicology
  • Lipoic acid binds covalently to provide a robust carboxylic acid surface
Spherical silver nanoparticles, 1 mg/ml (high concentration). Lipoic acid binds covalently to provide a robust carboxylic acid surface. BioPure materials are Sterile and Endotoxin to Free (<5 EU/ml). Precisely engineered, unagglomerated nanospheres with narrow size distributions (CVs <15%).
Each batch is extensively characterized to meet highest quality standards, ensuring reliable & reproducible results. All products supplied with COA's including batch-specific TEM, DLS, Zeta, UV-Vis measurements, etc.
Colloidal silver has applications in Diagnostics, Antimicrobial coatings, Electronics and Photovoltaics, SERS, Optics, Nanotoxicology, Plasmonics (SPR / Surface Plasmon Resonance), novel Color Enhancements and Pigments, Molecular Detection, Sensors, and Photothermal and Microwave-based Therapies.
Custom formulations and ISO13485/cGMP compliant materials can be provided on request.
Delivery information: Ships via FedEx Priority Overnight (Domestic). Cold packaging included.
Due to temperature-sensitive nature of these materials, they will only be shipped during the week in order to prevent products from sitting in an uncontrolled temperature environment over the weekend.
Caution: DO NOT FREEZE. Store at 2 to 8 °C and away from light when not in use to maximize shelf life. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty