Would you like to log in again?
NanoXact Aminated Silica-Shelled Gold Nanospheres 1 mg/ml Silica shell (high stability), nanoComposix
Supplier: nanoComposix
AUAH50-30M AUAH100-10M AUAH20-10M AUAH20-5M AUAH20-1M AUAH100-5M AUAH50-5M AUAH100-30M AUAH20-30M AUAH100-1M AUAH50-1M AUAH50-10M
103819-796EA 306.92 USD InStock
103819-796 103819-794 103819-788 103819-800 103819-786 103819-798 103819-804 103819-802 103819-792 103819-808 103819-806 103819-790
NanoXact Aminated Silica-Shelled Gold Nanospheres 1 mg/ml Silica shell (high stability), nanoComposix
Beads Microcarriers
Gold nanoparticles shelled with aminated silica, versatile surface for a wide variety of applications
  • High quality nanoparticles, precisely engineered and highly characterized
  • Unagglomerated and monodisperse
  • High concentration gold (1 mg/ml gold concentration)
  • Aminated silica shell offers robust and versatile surface for a wide variety of applications
Spherical gold nanoparticles, 1 mg/ml gold concentration. Aminated silica shell offers high stability in a variety of solvents and a versatile surface for attaching functional groups or creating hydrophobic and hydrophilic surfaces. Aminated silica shell allows easier deposition onto substrates and incororation into many different solvents, resins, and curable materials because the silica shell provents metal nanoparticles from touching directly / aggregating, helping to preserve the particles' nano and optical properties.
Each batch is extensively characterized to meet highest quality standards, ensuring reliable & reproducible results. All products supplied with batch-specific COA's with TEM, DLS, Zeta, & UV-Vis. to meet the highest quality standards
Colloidal gold has applications in Diagnostics, Lateral Flow Assays, Molecular Detection, Photothermal and Microwave-based Therapies, TEM labels, Nanotoxicology, Plasmonics (SPR / Surface Plasmon Resonance), and novel Color Enhancements and Pigments.
Custom formulations and ISO13485/cGMP compliant materials can be provided on request.
Delivery information: Ships via FedEx Priority Overnight (Domestic). Cold packaging included.
Due to temperature-sensitive nature of these materials, they will only be shipped during the week in order to prevent products from sitting in an uncontrolled temperature environment over the weekend.
Caution: DO NOT FREEZE. Store at 2 to 8 °C and away from light when not in use to maximize shelf life. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty