Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
BioPure Gold Nanospheres 1 mg/ml (Au) Carboxylic Acid surface (lipoic acid), nanoComposix
Supplier: nanoComposix
BIOPURE
AULB50-30M AULB100-30M AULB10-30M
103819-930EA 4027.69 USD InStock
103819-930 103819-924 103819-922
BioPure Gold Nanospheres 1 mg/ml (Au) Carboxylic Acid surface (lipoic acid), nanoComposix
Beads Microcarriers
Purified, concentrated, high quality gold nps, sterile and endotoxin-free. Carboxylic acid surface.
  • High quality nanoparticles, precisely engineered and highly characterized
  • Unagglomerated and monodisperse
  • High concentration materials (1 mg/ml, 5.08 mm, 1000 ppm)
  • Extensively purified to remove residual reactants
  • Manufactured sterile; provided in ultra to pure (Milli to Q) DI water
  • BioPure mateirals ideal for in to vitro and in to vivo experiments, nanotoxicology
  • Lipoic acid binds covalently to provide a robust carboxylic acid surface
Spherical gold nanoparticles, 1 mg/ml (high concentration), Lipoic acid binds covalently to provide a robust carboxylic acid surface. BioPure material is Sterile and Endotoxin to Free (<5 EU/ml). Precisely engineered, unagglomerated nanospheres with narrow size distributions (CVs <12% for 7 to 15 nm sizes, <15% for 20 to 100 nm sizes, and <18% for 5 nm size).
Each batch extensively characterized to meet highest quality standards, ensuring reliable & reproducible results. All products provided with batch-specific COA's with TEM, DLS, Zeta, & UV-Vis measurements.
Colloidal gold has applications in Diagnostics, Lateral Flow Assays, Molecular Detection, Photothermal and Microwave-based Therapies, TEM labels, Nanotoxicology, Plasmonics (SPR / Surface Plasmon Resonance), and novel Color Enhancements and Pigments.
Custom formulations and ISO13485/cGMP compliant materials can be provided on request.
Delivery information: Ships at ambient temperatures
Caution: DO NOT FREEZE. Store at 2 to 8 °C when not in use to maximize shelf life. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty