Would you like to log in again?
NanoXact Gold Nanospheres Dried Powder PVP (polyvinylpyrrolidone) excipient, nanoComposix
Supplier: nanoComposix
AUPD50-30MG AUPD10-30MG AUPD100-30MG
103819-964EA 1879.23 USD InStock
103819-964 103819-958 103819-956
NanoXact Gold Nanospheres Dried Powder PVP (polyvinylpyrrolidone) excipient, nanoComposix
Beads Microcarriers
High quality gold nanoparticles. Dried powder, redispersible in polar solvents (water, alcohols).
  • High quality nanoparticles, precisely engineered and highly characterized
  • Unagglomerated and monodisperse
  • Dried powder dispersible in a wide variety of polar and aqueous solvents
Spherical gold nanoparticles, Dried, PVP excipient to ensure particle stability so the materials redisperse without aggregation. PVP polymer surface provides strong steric stability (40 kD polyvinylpyrrolidone). Dried NanoXact materials are precisely engineered, unagglomerated nanospheres with narrow size distributions (CVs <12% for 7 to 15 nm sizes, <15% for 20 to 100 nm sizes, and <18% for 5 nm size). They are redispersible in a variety of polar solvents such as water, alcohols, DMF, and DMSO; medium dispersibility in chloroform, and low dispersibility in dichloromethane. Sonicate and vortex to re to suspend.
Each batch is extensively characterized to meet highest quality standards, ensuring reliable & reproducible results. All products supplied with batch-specific COA's with TEM images, TEM sizing, and UV-Vis.
Colloidal gold has applications in Diagnostics, Lateral Flow Assays, Molecular Detection, Photothermal and Microwave-based Therapies, TEM labels, Nanotoxicology, Plasmonics (SPR / Surface Plasmon Resonance), and novel Color Enhancements and Pigments.
Custom formulations and ISO13485/cGMP compliant materials can be provided on request.
Delivery information: Ships at ambient temperatures
Caution: DO NOT FREEZE. Store at 2 to 25 °C in a dry place. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty