Would you like to log in again?
BioPure Magnetite Nanoparticles 1 mg/ml PVP (polyvinylpyrrolidone), nanoComposix
Supplier: nanoComposix
MGPB20-5M MGPB20-10M MGPB20-1M MGPB20-30M
103820-378EA 306.92 USD InStock
103820-378 103820-380 103820-382 103820-376
BioPure Magnetite Nanoparticles 1 mg/ml PVP (polyvinylpyrrolidone), nanoComposix
Beads Microcarriers
Purified, high quality magnetite nanoparticles; sterile and endotoxin-free. PVP surface for stability.
  • High quality nanoparticles, precisely engineered and highly characterized
  • Unagglomerated and monodisperse
  • Extensively purified to remove residual reactants; manufactured sterile
  • Provided in ultra to pure (Milli to Q) DI water
  • BioPure matrial ideal for in to vitro and in to vivo experiments, nanotoxicology
  • Superparamagnetic properties at room temperature
Magnetite nanoparticles (Fe3O4), 20 nm, 1 mg/ml. PVP surface provides strong steric stability (40 kD PVP). BioPure materials are Sterile and Endotoxin to Free (<5 EU/ml). Precisely engineered and unagglomerated. These iron oxide particles exhibit superparamagnetic properties at ambient temperatures.
Each batch is extensively characterized to meet highest quality standards, ensuring reliable & reproducible results. All products supplied with batch-specific COA's with TEM, DLS, Zeta, & UV-Vis.
The size and superparamagnetic properties of these magnetite nanoparticles make them of interest for applications in many fields, including catalysis, biosensors, ferrofluids, magnetic separations and MRI contrast agents.
Custom formulations and ISO13485/cGMP compliant materials can be provided on request.
Delivery information: Ships via FedEx Priority Overnight (Domestic). Cold packaging included.
Due to temperature-sensitive nature of these materials, they will only be shipped during the week in order to prevent products from sitting in an uncontrolled temperature environment over the weekend.
Caution: DO NOT FREEZE. Store at 2 to 8 °C when not in use to maximize shelf life. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty