Would you like to log in again?
NanoXact Magnetite Nanoparticles Dried mg/ml PVP (polyvinylpyrrolidone), nanoComposix
Supplier: nanoComposix
MGPD20-500MG MGPD20-200MG MGPD20-100MG
103820-390EA 856.15 USD InStock
103820-390 103820-384 103820-386
NanoXact Magnetite Nanoparticles Dried mg/ml PVP (polyvinylpyrrolidone), nanoComposix
Beads Microcarriers
Concentrated high quality magnetite nanoparticles. Dispersible in polar solvents (water/alcohols).
  • High quality nanoparticles, precisely engineered and highly characterized
  • Unagglomerated and monodisperse
  • Superparamagnetic properties at room temperature
  • Dried nanomaterial can be re to dispersed in many polar solvents, such as water and alcohols
Magnetite nanoparticles (Fe3O4 Iron Oxide), 20 nm, Dried, PVP stabilized. Dried nanomaterials are dispersible in many polar solvents. These iron oxide particles are precisely engineered and unagglomerated, and exhibit superparamagnetic properties at ambient temperaturess. The size and superparamagnetic properties of these magnetite nanoparticles give them uses in a wide array of fields, such as catalysis, biosensors, ferrofluids, magnetic separation, and MRI contrast agents.
Each batch is extensively characterized to meet highest quality standards, ensuring reliable & reproducible results. All products supplied with batch-specific COA's with TEM, DLS, Zeta, & UV-Vis.
Custom formulations and ISO13485/cGMP compliant materials can be provided on request.
Delivery information: Ships at ambient temperatures
Caution: DO NOT FREEZE. Store at 2 to 25 °C in a dry place. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty