Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
NanoXact Aminated Mesoporous Silica Nanospheres Dried, nanoComposix
Supplier: nanoComposix
SHAD100-150MG SHAD100-25MG SHAD100-500MG
103820-548EA 1540 USD InStock
103820-548 103820-550 103820-552
NanoXact Aminated Mesoporous Silica Nanospheres Dried, nanoComposix
Beads Microcarriers
High quality mesoporous silica nanoparticles, provided with full COA
  • High quality nanoparticles, precisely engineered and highly characterized
  • Unagglomerated and monodisperse
  • Aminated silica surface offers robust and versatile surface for a wide variety of applications
Dried spherical mesoporous silica nanoparticles with an aminated surface. Like sponges, mesoporous silica nanoparticles (MSNs) can adsorb exceptionally large quantities of molecules in their interior spaces (pores) thanks to their high surface area. High quality mesoporous silica particles with uniform pore sizes and consistent pore structure can act as “advanced nanosponges” by only adsorbing molecules within a specific size range into their cavities. Uses for mesoporous silica range from catalysis and environmental chemical removal to biomedicine where they are widely used as drug delivery system (DDS). Their high surface area, easy surface modification, and biocompatibility make them the material of choice for targeted delivery of active pharmaceutical ingredients for cancer therapy and other therapies and treatments.
Each batch is extensively characterized to ensure it meets the highest quality standards so you can achieve reliable & reproducible results. Products are supplied with COA's that include batch-specific TEM, DLS, Zeta Potential, UV-Vis, & pH measurements.
Custom formulations and ISO13485/cGMP compliant materials can be provided on request.
Delivery information: Ships at ambient temperatures
Caution: DO NOT FREEZE. Store at 2 to 25 °C in a dry place. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty