Would you like to log in again?
Revco™ RDE Series –40 °C Ultra-Low Temperature Freezers, Thermo Scientific
Supplier: Thermo Scientific
Revco®
RDE50040LD RDE60040LA RDE50040FD RDE40040LV RDE60040LD RDE60040FV RDE50040FV RDE60040LV RDE40040LA RDE50040LA RDE50040FD RDE50040LA RDE60040LD RDE40040FD RDE50040LV RDE40040FA RDE40040FD RDE40040LD RDE60040LV RDE40040LD RDE60040FD RDE50040FV RDE60040FD RDE60040FV RDE40040FA RDE50040FA RDE40040LA RDE40040LV RDE60040LA RDE40040FV RDE60040FA RDE50040LD RDE50040FA RDE50040LV RDE40040FV RDE60040FA
76415-630EA 16369.98 USD InStock
76415-630 CA76415-634 CA76415-612 76415-646 76415-624 CA76415-636 CA76415-614 76415-626 76415-642 CA76415-630 76415-620 76415-644 76415-622 CA76415-632 76415-628 CA76415-616 CA76415-638 CA76415-618 76415-640 76415-636 CA76415-644 76415-614 CA76415-622 76415-638 CA76415-646 CA76415-624 76415-616 76415-632 CA76415-640 CA76415-620 76415-612 76415-634 CA76415-642 CA76415-626 76415-618 CA76415-628
Revco™ RDE Series –40 °C Ultra-Low Temperature Freezers, Thermo Scientific
Freezers Ultra Low Temperature Freezers
Revco™ RDE Series ‒40 °C ultra-low temperature freezers, powered by H-drive, feature three upright models, maximizing storage capacity from 40000 up to 60000 2 ml vials.
  • Systems save up to almost 50% in energy usage
  • Smart-Vue compatible
  • Easy to read temperature with simple capacitive touch buttons
  • Accommodate a variety of storage needs and lab footprint specifications
  • Security features including alarm settings and padlock-compatible doors
Natural hydrocarbon refrigerants reduce system operating stresses promoting outstanding reliability and worry-free sample protection. New ergonomically designed handle, high contrast eye-level display, and comfortable easy-access shelf locations for greater convenience.
Certifications: cULus certified.
The models with LA at the end of the model name are also FDA Class II Compliant.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty