Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Blue Phantom® Internal Jugular Central Line Ultrasound Trainers
Supplier: CAE HEALTHCARE, INC. SE
Blue Phantom®
BPIJ500 BPIJ500-C
470327-444EA 2510.79 USD InStock
470327-444 470327-396
Blue Phantom® Internal Jugular Central Line Ultrasound Trainers
Models Human Biology Models
Compact and lightweight.
  • Can be used with any ultrasound system
  • Self-healing and ultra-durable tissue
  • Simulate arterial pulsation via the included hand bulb
  • Available in transparent or opaque material
The internal jugular central line ultrasound simulator offers superb internal and external anatomical realism. Developed using the innovative Life Cast™ modeling techniques, the external anatomy is presented in tremendous detail and the internal anatomy matches human anatomy, allowing you to practice and teach on this extremely lifelike central line manikin. The specially designed tissues will never clog needles and will always image like human tissue without unrealistic ultrasound system settings.
The lightweight and portable internal jugular training manikin contains vascular anatomy including the internal jugular vein (IJ) and the carotid artery. Internal structures also include the trachea and clavicle providing you with superb realism – using any ultrasound system equipped with a vascular access ultrasound transducer. External landmarks also include a palpable suprasternal notch, trachea, and clavicle. This central venous access ultrasound phantom is specifically designed for teaching and practicing internal jugular (IJ) cannulation; the training model accommodates both ultrasound guidance and blind insertion technique.
Ordering information: These items cannot be returned if used. Please consult with your sales representitive before purchasing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty