Would you like to log in again?
Blue Phantom® TTE and Peri cardiocentesis Ultrasound Trainer
Supplier: CAE HEALTHCARE, INC. SE
Blue Phantom®
BPTTE1701-HEAD BPP-022
470327-446EA 27369.59 USD InStock
470327-446 470327-460
Blue Phantom® TTE and Peri cardiocentesis Ultrasound Trainer
Models Human Biology Models
Extremely realistic imaging characteristics.
  • Acoustic characteristics of real human tissue
  • Feels and cannulates like real human tissue
  • Life size ultrasound training model
  • Heart contains accurate anatomy
  • Usable from all appropriate imaging positions
  • Excellent for 2-D, 3-D, and 4-D training
  • Extremely durable
Excellent for training clinicians in the techniques associated with transthoracic echocardiography (TTE), and ultrasound guided pericardiocentesis procedural training. Blue Phantom®’s cardiac echo ultrasound training platform allows users to develop and practice ultrasound imaging skills as transthoracic echocardiography and ultrasound guided pericardiocentesis.
These skills include; using ultrasound system controls, transducer placement, positioning and movement, applying proper transducer pressure to obtain images, recognition of the cardiac structures, lung and rib artifacts and other anatomical structures and guiding needles and catheters to the pericardial space for pericardiocentesis training. Offers an extremely realistic imaging training platform as you utilize your own ultrasound system for simulation training. Self-healing simulated tissue allows for repeated procedural training without fluid leakage.
This extremely realistic life size ultrasound simulation training model extends from the neck to the mid thigh. This simulation trainer will perform well using any ultrasound imaging system configured with the appropriate transducer for echocardiography imaging procedures. The uncompromising quality allows clinicians to utilize the model and repeatedly practice the procedure without the high costs of replacing disposable parts.
Users can expect extreme durability with the simulated tissue able to perform well for thousands of procedures. This ultrasound simulation model is excellent for cardiology, anesthesiology, emergency medicine, ultrasound training programs, simulation centers, surgical skills centers, medical education facilities, and ultrasound manufacturers for ultrasound education and demonstrations.
Ordering information: These items cannot be returned if used. Please consult with your sales representitive before purchasing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty