Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cell Systems Human Brain Pericytes, Primary Cell and Media Company

Supplier: Cell Systems
ACBRI498
103810-660EA 1047.69 USD InStock
103810-660
Cell Systems Human Brain Pericytes, Primary Cell and Media Company
Cells
Our primary human cells offer unmatched consistency and a more biologically relevant tool than other cell types. Our cells are antibody-free and come from a deep inventory to help researchers with reproducibility and improved human insights.

  • Highly proliferative and healthy (~85% viability)
  • Antibody free and more biologically relevant
  • Offered with high cell count (~1 million cells per vial)
  • Well-cited in scientific journals
  • Ideal for 2D and 3D cultures, microfluidic devices and co-cultures

Primary Human Brain Pericytes (ACBRI498) were isolated by enzymatic dissociation of normal human cerebral cortex tissue. This cell is antibody free. Each vial will initiate a 25 to 75 cm² cell culture. These primary cells are isolated, frozen and qualified using Cell Systems Certified Media and Reagents--which are specified for use in the research environment. Cell Systems primary cells are exclusively from normal, site-specific human tissue specimen, and available in frozen vial or proliferating flask.

These cells were originated using Cell Systems Complete Serum-Free Medium (SF-4Z0-500), and subsequently grown and passaged in Cell Systems Complete Medium (4Z0-500). They are available at Passage 3 (< 12 cumulative population doublings) cryopreserved in Cell Systems Cell Freezing Medium™ (4Z0-705). This vial will initiate a Passage 4 cell culture in a 75 cm² flask. Each vial contains more than 1×106 cells.

Each vial of cells is shipped with vials of Bac-Off® (antibiotic) and CultureBoost™ (animal derived growth factors) or CultureBoost-R™ (human recombinant growth factors) at no additional cost.

Standard Tests:
HIV Serologic Test (donor level HIV AB EIA) -Negative
RPR Syphilis Test - Negative
Hepatitis B (HBV) PCR Test (frozen cells) - Negative
Hepatitis C (HCV) PCR Test (frozen cells) - Negative
Mycoplasma spp. (frozen cells) (ATCC method by CLIA Licensed Clinical Lab) - Negative.

Characterization:
Cytoplasmic alpha-Actinin Filaments > 95% positive by immunofluorescence
Cytoplasmic Desmin Intermediate Filament > 95% positive by immunofluorescence
Cytoplasmic uptake of Di-L-Ac-LDL < 1% positive by immunofluorescence
CD13 pericyte marker > 98% positive byence
a-SMA pericyte marker > 98% positive by immunofluorescence
CD31 endothelial cell marker < 2% positive by immunofluorescence
Uptake of Dil-Ac-LDL < 2% positive by immunofluorescence
MAP2 neuronal marker < 2% positive by immunofluorescence
Neurofilament neuronal marker < 2% positive by immunofluorescence
S100A4 fibroblast marker < 2% positive by immunofluorescence
GFAP astrocyte marker < 2% positive by immunofluorescence
GS astrocyte marker < 2% positive by immunofluorescence
CD11b microglial marker < 2% positive by immunofluorescence
Ibal microglial marker < 2% positive by immunofluorescence.

Caution: For Research Use Only

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty