Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Rabbit Polyclonal Antibody (Gold, 25nm)

Supplier: Electron Microscopy Sciences
25355 25356
100487-742EA 698.42 USD InStock
100487-742 100487-744
Anti-IgG Rabbit Polyclonal Antibody (Gold, 25nm)
Antibodies
The Conventional Immunogold Reagents are the classical conjugates in immuno electron microscopy; they are a good choice when the antigen is abundant and the accessibility of the antigen is relatively good.

Conventional ImmunoGold Reagents are monodisperse size population makes the conjugates suited for multiple labeling with no overlap. The Conventional Immunogold Reagents are the classical conjugates in immuno electron microscopy; they are a good choice when the antigen is abundant and the accessibility of the antigen is relatively good. The conventional labeling approach in transmission and scanning electron microscopy utilizes secondary immunogold reagents based on particles that can be observed without enhancement. The particle population is monodisperse and thus shows minimal size variation and overlap.The reagents are supplied in PBS with 1% Bovine Serum Albumin and 15 mM NaN3.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Gold, 25nm
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Sheep

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty