Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

IDO-IN-2 ≥98% (by HPLC)

Synonyms: 1-(4-(Diisobutylamino)-2'-(2H-tetrazol-5-yl)-[1,1'-biphenyl]-3-yl)-3-(p-tolyl)urea

AB287066-25MG AB287066-5MG
76411-826EA 1075.95 USD InStock
76411-826 76411-828
IDO-IN-2 ≥98% (by HPLC)
IDO-IN-2
IDO-IN-2 is a potent indoleamine 2,3-dioxygenase (IDO) inhibitor (IC₅₀ = 0.068 µM).

  • A potent indoleamine 2,3-dioxygenase (IDO) inhibitor


Formula: C₂₉H₃₅N₇O
MW: 497.65 g/mol
Storage Temperature: Freezer
MDL Number: MFCD30728441
CAS Number: 1668565-74-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty