Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

CH7057288 ≥98% (by HPLC)

Supplier: BioVision

AB287079-5MG AB287079-25MG
76411-898EA 1307.02 USD InStock
76411-898 76411-900
CH7057288 ≥98% (by HPLC)
CH7057288
CH7057288 as a potent and selective TRK (tropomyosin receptor kinase) inhibitor that may be useful in inhibiting TRK fusion-positive cancer cell growth. CH7057288 shows selective inhibitory activity against TRKA, TRKB, and TRKC in cell-free kinase assays and suppresses proliferation of TRK fusion–positive cell lines, but not that of TRK-negative cell lines.

  • A selective tropomyosin receptor kinase (TRK) inhibitor


Formula: C₃₂H₃₁N₃O₅S
MW: 569.68 g/mol
Storage Temperature: Freezer
CAS Number: 2095616-82-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty