Would you like to log in again?
Muramyl Dipeptide
Supplier: BioVision
AB287084-5MG AB287084-1MG
76411-964EA 151.66 USD InStock
76411-964 76411-966
Muramyl Dipeptide
Proteins and Peptides
Muramyl dipeptide is a synthetic immunoreactive peptide that activates NOD2 receptors. Induces polarization of proinflammatory Ly6Chigh into Ly6Clow patrolling monocytes. Also inhibits HIV replication in CD4⁺ H9 lymphocytes.
  • NOD2 receptors activator
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty