Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

TCE35031 ≥98% (by HPLC)

Supplier: BioVision
Synonyms: 2-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)oxy)acetic acid

AB287105-25MG AB287105-5MG
76414-590EA 151.66 USD InStock
76414-590 76414-588
TCE35031 ≥98% (by HPLC)
TCE35031
TCE35031 is an analog of Thalidomide (Cat. No. 2020) commonly used as a precursor to PROTACs (proteolysis-targeting chimera technologies) that hijack cereblon as the E3 ubiquitin ligase component. Supplied with a carboxylic acid functional handle at a position known not to significantly affect binding to cereblon, for ready conjugation to a linker/target protein ligand.

  • A cereblon ligand for PROTACs


Formula: C₁₅H₁₂N₂O₇
MW: 332.26 g/mol
Storage Temperature: Freezer
MDL Number: MFCD31560477
CAS Number: 1061605-21-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty