Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

E7820 ≥98% (by HPLC)

Supplier: BioVision
Synonyms: 3-Cyano-N-(3-cyano-4-methyl-1H-indol-7-yl)benzenesulfonamide3-Cyano-N-(3-cyano-4-methyl-1H-indol-7-yl)benzenesulphonamide

AB287106-25MG AB287106-5MG
76414-594EA 151.66 USD InStock
76414-594 76414-592
E7820 ≥98% (by HPLC)
E7820
E7820 is a small molecule sulfonamide derivative with potential antiangiogenic and antitumor activities. E7820 inhibits angiogenesis by suppressing integrin alpha 2, a cell adhesion molecule expressed on endothelial cells. Inhibition of integrin alpha 2 leads to an inhibition of cell-cell interactions, endothelial cell-matrix interactions, vascular endothelial cell proliferation and angiogenesis.

  • An angiogenesis inhibitor


Formula: C₁₇H₁₂N₄O₂S
MW: 336.37 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09833234
CAS Number: 289483-69-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty