Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

AEM1 ≥98% (by HPLC)

Supplier: BioVision

AB287111-1MG AB287111-5MG
76414-638EA 556.03 USD InStock
76414-638 76414-636
AEM1 ≥98% (by HPLC)
AEM1
AEM1 is an inhibitor of Nrf2-induced gene transcription at the antioxidant response element in cells at 1 to 10 µM. It sensitizes lung carcinoma A549 cells to chemotherapeutic agents and inhibits the growth of A549 cells in vitro and in vivo in animals.

  • An inhibitor of Nrf2-induced gene transcription


Formula: C₂₀H₁₄FN₃O₂S
MW: 379.41 g/mol
Storage Temperature: Freezer
CAS Number: 1030123-90-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty