Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PF-3758309 ≥98% (by HPLC)

Danger
Synonyms: (S)-N-(2-(Dimethylamino)-1-phenylethyl)-6,6-dimethyl-3-((2-methylthieno[3,2-d]pyrimidin-4-yl)amino)-4,6-dihydropyrrolo[3,4-c]pyrazole-5(1H)-carboxamide

AB287116-5MG AB287116-25MG
76408-174EA 231.06 USD InStock
76408-174 76408-172
PF-3758309 ≥98% (by HPLC)
PF-3758309
PF-3758309 is a potent small molecule inhibitor of p21-activated kinase 4 (PAK4) with potential antineoplastic activity. PF-3758309 binds to PAK4, inhibiting PAK4 activity and cancer cell growth. PAK4, a serine/threonine kinase belonging to the p21-activated kinase (PAK) family, is often upregulated in a variety of cancer cell types and plays an important role in cancer cell motility, proliferation, and survival.

  • A p21-activated kinase 4 (PAK4) inhibitor


Formula: C₂₅H₃₀N₈OS
MW: 490.62 g/mol
Storage Temperature: Freezer
MDL Number: MFCD18633226
CAS Number: 898044-15-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty