Would you like to log in again?
Isolation of Human Cell, Lymphoprep™, BioVision
Supplier: BioVision
AB286892-500ML M1274-4PK AB286892-250ML
76415-188EA 470.62 USD InStock
76415-188 76415-184 76415-186
Isolation of Human Cell, Lymphoprep™, BioVision
Cell Separation Media
Used in isolation of human mononuclear cells.
Lymphoprep™ is a ready-made, sterile and endotoxin tested density gradient medium recommended for the isolation of pure lymphocyte suspensions for tissue typing, anti-lymphocyte sera and immunological research. The solution contains sodium diatrizoate (9.1% (w/v) and polysaccharide (5.7% (w/v).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty