Would you like to log in again?
Magnetic Separation Rack, BioVision
Supplier: BioVision
AB286900-1UNITS
76417-110EA 948.11 USD InStock
76417-110
Magnetic Separation Rack, BioVision
Racks Magnetic Separation Racks
A rack to separate magnetic beads in tubes.
BioVision’s Magnetic Separation Rack is used to capture and separate magnetic beads used in magnetic beads based purification techniques. The rack captures magnetic beads which may be bound to DNA, viruses or cells in tubes. The biomolecules or cells are eluted and concentrated from the magnetic beads using appropriate buffers for downstream applications. BioVision’s Magnetic Rack uses permanent magnets and has a compact design to accommodate different sizes of tubes including twelve 1.5 ml tubes, two 15 ml tubes and two 50 ml tubes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty