Would you like to log in again?
Social Distance Slip-Gard™ Floor Signs; 6' (Arrows/Footprints), Accuform®
Supplier: Accuform
MFS348 MFS346
76416-896EA 40.91 USD InStock
76416-896 76416-898
Social Distance Slip-Gard™ Floor Signs; 6' (Arrows/Footprints), Accuform®
Signs
Floor signs alert and remind those to practice social and physical distancing to prevent the spread of COVID-19, the Coronavirus. 
  • Constructed of thick adhesive vinyl
  • Non-slip, pebble-textured surface provides added durability
  • Strong, permanent, acrylic adhesive back with peel-off liner
  • Adheres firmly to smooth, clean, dry surfaces
Subsurface is printed and laminated to protect the graphics and message from heavy pedestrian traffic.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty