Would you like to log in again?
Social Distance Signs: We Appreciate Your Patience When Lining Up to Pay, Accuform®
Supplier: Accuform
MGNG526VP MGNG524VP MGNG526VA MGNG524VS MGNG524VA MGNG526VS
76416-996EA 13.81 USD InStock
76416-996 76417-002 76417-004 76417-006 76416-998 76417-000
Social Distance Signs: We Appreciate Your Patience When Lining Up to Pay, Accuform®
Signs
Signs alert and remind those to practice social and physical distancing to prevent the spread of COVID-19, the Coronavirus. 
  • Multiple material options are available
Adhesive Vinyl signs feature a printed matte surface with permanent acrylic adhesive back. Vinyl signs are moisture-resistant for a variety of light-duty indoor and sheltered outdoor marking needs.
Plastic signs are durable with slight flex capabilities. Plastic is a good material choice for indoor and sheltered outdoor environments. The printed surface has a clear UV protective overcoat.
Aluminum signs provide the strength of metal without rusting. Their printed surface is protected by a UV coating to withstand splashes, wash-downs, and outdoor conditions, plus abrasion and chemical resistance.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty