Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Social Distance Signs: Please Wait Here Until Called to Step Forward, Accuform®
Supplier: Accuform
MGNG537VA MGNG537VS MGNG535VP MGNG535VA MGNG537VP MGNG535VS
76417-036EA 21.59 USD InStock
76417-036 76417-038 76417-040 76417-032 76417-042 76417-034
Social Distance Signs: Please Wait Here Until Called to Step Forward, Accuform®
Signs
Signs alert and remind those to practice social and physical distancing to prevent the spread of COVID-19, the Coronavirus. 
  • Multiple material options are available
Adhesive Vinyl signs feature a printed matte surface with permanent acrylic adhesive back. Vinyl signs are moisture-resistant for a variety of light-duty indoor and sheltered outdoor marking needs.
Plastic signs are durable with slight flex capabilities. Plastic is a good material choice for indoor and sheltered outdoor environments. The printed surface has a clear UV protective overcoat.
Aluminum signs provide the strength of metal without rusting. Their printed surface is protected by a UV coating to withstand splashes, wash-downs, and outdoor conditions, plus abrasion and chemical resistance.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty