Would you like to log in again?
Lentivirus and Retrovirus Purification Kit, BioVision
Supplier: BioVision
AB288104-100 AB288104-20TEST
76416-416EA 1127.2 USD InStock
76416-416 76416-418
Lentivirus and Retrovirus Purification Kit, BioVision
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Magnetic beads based kits
  • Does not need ultracentrifugation
  • High yield and fast concentration (2 to 1000X)
  • Ideal for cell culture transduction or infection
  • Purify viruses in 30 to 45 min
BioVision’s Lentivirus & Retrovirus Purification Kit is used to capture, concentrate and store lentiviruses and retroviruses using magnetic beads. Viruses are important gene delivery tools, which have been used in gene therapy and RNAi delivery. Traditionally, viruses are purified by ultracentrifugation using CsCl, which is time consuming and is limited to the volume of cell lysate to be processed. BioVision’s Purification kit uses magnetic nanoparticles to capture viruses by electrostatic and hydrophobic interactions with 80 to 99% efficiency and the purification process does not need ultracentrifugation. The viruses captured on Lentivirus (LV)-Retrovirus (RV)-magnetic beads can be used directly for multiple assays. Alternatively, viruses can be eluted with the elution buffer and used for downstream applications.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty