Would you like to log in again?
Coronavirus Rapid RT-qPCR Detection Kit, BioVision
Supplier: BioVision
AB287868-96TEST
76416-424EA 806.63 USD InStock
76416-424
Coronavirus Rapid RT-qPCR Detection Kit, BioVision
Assays qPCR Assays
qPCR based detection kit to detect SARS-CoV-2.
  • Compatible with standard RT-qPCR machines (Bio-Rad CFX96 QuantStudio’s 7 Flex system)
  • Highly specific for the ORF1ab and N target markers recommended by WHO and US CDC
  • Includes a positive control
  • Reliable and Ready-to-use
  • Results ready in less than 2 hours
BioVision’s Coronavirus rapid RT-qPCR Detection Kit is used for the detection of SARS-CoV-2 in respiratory specimens using Reverse Transcriptase Quantitative Polymerase Chain Reaction (RT-qPCR). It allows the efficient cDNA synthesis and qPCR in a single tube. This kit includes a 2X RT-qPCR Mastermix that contains all the reagents supplied in a 2X concentration to perform the qPCR. A separate RT-qPCR Enzyme Mix for cDNA synthesis is included the kit. Additionally, the kit contains 2 vials of primers and probes specific for the ORF1ab and N target. The PCR Detection Kit will facilitate the Coronavirus research and improve patient treatment.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty