Would you like to log in again?
Viral RNA Extraction Kit for Respiratory Specimens, BioVision
Supplier: BioVision
AB288110-250 AB288110-50TEST
76416-428EA 955.24 USD InStock
76416-428 76416-426
Viral RNA Extraction Kit for Respiratory Specimens, BioVision
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Using spin columns
  • Rapid, easy and convenient
  • Extract Viral RNA for many downstream applications
  • High quality spin columns
  • Highly pure, high yield
BioVision’s Viral RNA Extraction Kit for Respiratory Specimens provides an easy and reliable method for isolating total viral RNA from plasma, serum, nasopharyngeal or oropharyngeal aspirates or washes, nasopharyngeal or oropharyngeal swabs, broncheoalveolar lavage, tracheal aspirates and sputum using spin columns. This procedure has been tested for isolating nucleic acids from Hepatitis A, Hepatitis C and HIV. The isolated RNA can be used for PCR, qRT-PCR and other downstream applications. BioVision offers the Viral RNA Extraction Kit to facilitate research on Viruses.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty