Would you like to log in again?
Homocysteine Lyase Activity Assay Kit (Fluorometric), BioVision
Supplier: BioVision
AB284529-100TEST
76415-398EA 1054.3 USD InStock
76415-398
Homocysteine Lyase Activity Assay Kit (Fluorometric), BioVision
Assays Immunological Assays
To assay HCYase activity in bacterial samples
  • Sensitive method to assay HCYase activity in bacterial samples
  • Quick
  • Simple
Homocysteine α,γ-Lyase (HCYase) is a pyridoxal 5'-phosphate dependent enzyme that catalyzes the breakdown of homocysteine to hydrogen sulfide, ammonia, and α-ketobutyrate. HCYase is present in high activity in the anaerobic protozoan parasite Trichomonas vaginalis but is absent from mammals. The Trichomonas HCYase is highly selective for homocysteine as compared to cysteine. Additionally, it has broad pH stability (pH 6-8) and is thermally stable up to 50°C. This makes rHCYase a promising candidate for therapeutic applications including the diagnosis of hyperhomocystenemia, which has been demonstrated to be a major risk factor in cardiovascular diseases. BioVision’s Homocysteine Lyase Activity Assay Kit is a simple, quick and sensitive method to assay HCYase activity in bacterial samples. The assay relies on the HCYase-catalyzed release of hydrogen sulfide (H2S), which reacts with a fluorogenic probe to form a stable fluorophore measured at Ex/Em = 368/460 nm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty