Would you like to log in again?
NADH Oxidase Activity Assay Kit (Colorimetric), BioVision
Supplier: BioVision
AB284530-100TEST
76415-402EA 788.15 USD InStock
76415-402
NADH Oxidase Activity Assay Kit (Colorimetric), BioVision
Assays Enzyme Assays
to determine the NADH Oxidase activity in a variety of sample types
  • Used to determine NOX activity in a variety of sample types with a detection limit of ~ 1 mU of NOX activity per reaction
  • Simple
  • Rapid
NADH oxidase (Nicotinamide Adenine Dinucleotide Phosphate Oxidase, NOX) is a flavoprotein that catalyzes the oxidation of NADH by molecular oxygen to yield NAD+. NOX is one of the well-known cancer biomarkers and is shown to be stimulated by growth factors and hormones. Several studies revealed the high activity of NADH oxidase in cancer patients as compared to the healthy volunteers. More importantly, inhibition of NOX correlates with the reduced cancer cell growth and induction of apoptosis. RNA interference targeting NOX expression in cancer cells effectively restored the non-cancerous phenotypes, further supporting the vital role of NOX in cancer progression. Furthermore, NOX is critical for the regeneration of NAD+, for use in glycolysis and for the reduction of oxygen thereby preventing the formation of damaging reactive oxygen species. This makes NADH oxidase an attractive therapeutic target. BioVision’s NADH Oxidase Activity Assay Kit couples the oxidation of NADH by NOX and the reduction of a colored substrate to a colorless product. This result in the decrease of absorbance measured at 600 nm. The kit can be used to determine NOX activity in a variety of sample types with a detection limit of ~ 1 mU of NOX activity per reaction.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty