Would you like to log in again?
Methionine Adenosyltransferase Activity Assay Kit (Colorimetric), BioVision
Supplier: BioVision
AB284535-100TEST
76416-460EA 1011.79 USD InStock
76416-460
Methionine Adenosyltransferase Activity Assay Kit (Colorimetric), BioVision
Assays Enzyme Assays
Measurement of MAT activity in complex biological matrices
  • Does not require complicated sample processing
  • Limit of quantification of 2 mU MAT activity per well
  • Simple to perform
  • High-throughput adaptable
  • Homogeneous
Methionine Adenosyltransferases (MATs, EC 2.5.1.6), also known as Adenosylmethionine Synthetases are a family of enzymes that synthesize S-adenosyl-L-methionine (SAMe) from L-methionine and ATP. SAMe is a primary biochemical alkylation agent (one of only two possible methyl group donors) and is also a vital metabolic precursor of the transsulfuration and polyamine synthesis (aminopropylation) pathways. All organisms express at least one MAT enzyme. In mammals, three isozymes of MAT have been identified. MAT1 and MAT3 isozymes are predominantly expressed in the liver, whereas MAT2A is expressed in most tissues. Upregulation of the MAT2A isozyme has been linked to several human diseases. MAT2A has become a popular drug target for novel cancer therapeutics as well as for hepatic fibrosis and non-alcoholic fatty liver disease. BioVision’s Methionine Adenosyltransferase Activity Assay Kit enables the rapid measurement of MAT activity in complex biological matrices. The assay is based on the detection of pyrophosphate, which is generated stoichiometrically during the generation of SAMe. Pyrophosphate is enzymatically metabolized to an intermediate product, which reacts with the probe to form a stable chromophore that is detected by absorbance at 570 nm. The assay is homogeneous, simple to perform and does not require complicated sample processing. The assay is high-throughput adaptable and has a limit of quantification of 2 mU MAT activity per well.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty