Would you like to log in again?
Sucrose Phosphorylase Activity Assay Kit (Colorimetric), BioVision
Supplier: BioVision
AB284536-100TEST
76416-462EA 969.37 USD InStock
76416-462
Sucrose Phosphorylase Activity Assay Kit (Colorimetric), BioVision
Assays Enzyme Assays
To determine the activity of Sucrose Phosphorylase
  • Rapid
  • Can detect as low as 10 mU under the assay conditions
  • Sensitive
  • Convenient for measuring Sucrose Phosphorylase activity
Sucrose Phosphorylase (EC 2.4.1.7) is a transglucosidase enzyme, which plays a key role in the metabolism of sucrose, starch as well as other metabolites. It catalyzes the conversion of sucrose to α-D-glucose-1-phosphate (G1P) and D-fructose. It is present in both gram positive and negative bacteria such as in Leuconostoc mesenteroides, Bifidobacterium longum, Pseudomonas sacclwrophila and Lactobacillus reuteri. Sucrose Phosphorylase is used as a biocatalyst to produce fructose syrup in food industry and for determining inorganic phosphate in clinical analysis. BioVision’s Sucrose Phosphorylase Activity Assay Kit can be used to determine the activity of Sucrose Phosphorylase in different microorganisms. The assay converts Sucrose to G1P, which is then detected by a set of enzymatic reactions to generate a colored product. The colored product can be measured by absorbance (OD 450 nm) and is directly proportional to the Sucrose Phosphorylase activity. BioVision’s Sucrose Phosphorylase Activity Assay Kit is a rapid, sensitive and a convenient tool for measuring Sucrose Phosphorylase activity. It can detect as low as 10 mU under the assay conditions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty